siRNA Details
Gene Symbol: Zfp672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028331.2 | 2 | 121 | |||
NR_046280.1 | 2 | 121 | ||||
GenBank mRNA | AK008471.1 | - | 2 | 97 | ||
AK019676.1 | - | 2 | 71 | |||
AK029711.1 | - | 2 | 289 | |||
AK085166.1 | - | 2 | 81 | |||
BB615378.1 | - | 2 | 282 | |||
BC034706.1 | - | 2 | 68 | 3582315 | ||
BG091612.1 | - | 2 | 82 | |||
BU148968.1 | - | 2 | 53 | |||
BY017375.1 | - | 2 | 102 | |||
BY715308.1 | - | 2 | 71 | |||
CA321162.1 | - | 2 | 54 | |||
CV783882.1 | - | 2 | 70 | |||
DV654279.1 | - | 2 | 155 |
