siRNA Details
Gene Symbol: 4930429B21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027966.1 | 1 | 1810 | |||
GenBank mRNA | AK018559.1 | - | 1 | 1526 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_026249.1 | - | 1526 | Chr. - - [-] on Build |
