siRNA Details
Gene Symbol: 4930442L01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015596.1 | 2 | 610 | |||
GenBank mRNA | AK006795.1 | - | 1 | 785 | ||
AK015364.1 | - | 2 | 610 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706776.1 | - | 1 | 785 | Chr. 3 - 96831058 - 96832050 [-] on Build GRCm38 |
