siRNA Details
Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024496.1 | 2 | 714 | |||
GenBank mRNA | BC042539.1 | - | 2 | 714 |
Gene Symbol: LOC105379526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X97876.1 | - | 3 | 1001 |
