siRNA Details
Gene Symbol: LOC100130872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024569.1 | 2 | 499 | |||
GenBank mRNA | CR627052.1 | - | 2 | 499 |
Gene Symbol: SPON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB104725.1 | - | 1 | 90 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB184721.1 | - | 1 | 84 | Chr. 4 - 1172049 - 1205088 [-] on Build GRCh38 |
