siRNA Details
Gene Symbol: LINC00305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027245.1 | 4 | 566 | |||
GenBank mRNA | AY336744.1 | - | 3 | 329 | ||
BC029565.1 | - | 4 | 570 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG771578.1 | - | 4 | 575 | Chr. 18 - 64080051 - 64149030 [-] on Build GRCh38 |
