siRNA Details
Gene Symbol: LINC00305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027245.1 | 4 | 615 | |||
GenBank mRNA | BC029565.1 | - | 4 | 619 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG771578.1 | - | 4 | 624 | Chr. 18 - 64080051 - 64149030 [-] on Build GRCh38 |
