siRNA Details
Gene Symbol: VLDLR-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015375.2 | 3 | 512 | |||
GenBank mRNA | AF424541.1 | - | 7 | 977 | ||
BC004474.2 | - | 3 | 512 | |||
BE086096.1 | - | 3 | 308 | |||
DA119626.1 | - | 3 | 449 |
