siRNA Details
Gene Symbol: CROCCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023386.1 | 15 | 3308 | |||
GenBank mRNA | AB067509.1 | - | 15 | 3307 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CROCCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023386.1 | 15 | 3308 | |||
GenBank mRNA | AB067509.1 | - | 15 | 3307 |