siRNA Details
Gene Symbol: OLMALINC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026762.1 | 1 | 336 | |||
GenBank mRNA | AK023036.1 | - | 1 | 69 | ||
BC024924.1 | - | 1 | 297 | |||
BC045787.1 | - | 1 | 315 | |||
CB159470.1 | - | 1 | 363 |
