siRNA Details
Gene Symbol: AFG3L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003226.1 | 8 | 912 | |||
NR_003227.1 | 7 | 739 | ||||
NR_003228.1 | 8 | 878 | ||||
GenBank mRNA | AF329691.1 | - | 7 | 663 | ||
AF329692.1 | - | 2 | 214 | |||
AK056488.1 | - | 8 | 883 | |||
AK091293.1 | - | 8 | 875 | |||
AK097044.1 | - | 7 | 726 | |||
AK126911.1 | - | 6 | 703 | |||
AK128352.1 | - | 1 | 578 | |||
AL832984.1 | - | 2 | 136 | |||
BC021245.2 | - | 6 | 485 |
