siRNA Details
Gene Symbol: ZNF674-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015378.3 | 2 | 1115 | |||
GenBank mRNA | AK056314.1 | - | 2 | 1086 | ||
BC034616.1 | - | 2 | 1112 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD639292.1 | - | 3 | 237 | Chr. X - 46545509 - 46547861 [+] on Build GRCh38 |
