siRNA Details
Gene Symbol: LINC00698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027104.1 | 7 | 960 | |||
GenBank mRNA | BC039502.1 | - | 6 | 552 | ||
BC043407.1 | - | 9 | 1081 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM904732.1 | - | 6 | 551 | Chr. 3 - 62950459 - 63124605 [+] on Build GRCh38 |
