siRNA Details
Gene Symbol: ANXA2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001446.2 | 1 | 52 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ANXA2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001446.2 | 1 | 52 |