siRNA Details
Gene Symbol: TPM3P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003148.3 | 2 | 361 | |||
GenBank mRNA | BC061910.1 | - | 1 | 172 | ||
BX647962.1 | - | 2 | 348 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TPM3P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003148.3 | 2 | 361 | |||
GenBank mRNA | BC061910.1 | - | 1 | 172 | ||
BX647962.1 | - | 2 | 348 |