siRNA Details
Gene Symbol: TTTY10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001542.1 | 1 | 939 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF332239.3 | - | 1 | 939 | Chr. Y - 20465668 - 20519228 [-] on Build GRCh38 |
