siRNA Details
Gene Symbol: DNM1P46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003260.1 | 2 | 231 | |||
GenBank mRNA | AK127834.1 | - | 2 | 231 | ||
BC101080.2 | - | 2 | 119 | |||
BC101081.2 | - | 2 | 119 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DNM1P46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003260.1 | 2 | 231 | |||
GenBank mRNA | AK127834.1 | - | 2 | 231 | ||
BC101080.2 | - | 2 | 119 | |||
BC101081.2 | - | 2 | 119 |