siRNA Details
Gene Symbol: HIGD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002780.1 | 3 | 759 | |||
GenBank mRNA | BC040890.1 | - | 3 | 759 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM564128.1 | - | 3 | 779 | Chr. 15 - 72675889 - 72686150 [-] on Build GRCh38 |
