siRNA Details
Gene Symbol: DUBR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028301.1 | 1 | 272 | |||
NR_028302.1 | 1 | 272 | ||||
GenBank mRNA | AK055934.1 | - | 1 | 140 | ||
AK309100.1 | - | 1 | 272 | |||
BC039550.1 | - | 1 | 239 | |||
DA339707.1 | - | 1 | 272 |
