siRNA Details
Gene Symbol: SERTAD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024337.1 | 1 | 148 | |||
GenBank mRNA | BC066352.1 | - | 1 | 147 | ||
BC094794.1 | - | 1 | 184 | |||
BP422084.1 | - | 1 | 60 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG709605.1 | - | 1 | 150 | Chr. 1 - 210231479 - 210234124 [-] on Build GRCh38 |
