siRNA Details
Gene Symbol: MIR9-3HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015411.1 | 7 | 3671 | |||
NR_133001.1 | 7 | 4251 | ||||
NR_133002.1 | 6 | 3552 | ||||
NR_133003.1 | 7 | 3586 | ||||
GenBank mRNA | AK054710.1 | - | 1 | 1287 | ||
AK096359.1 | - | 1 | 1786 |
