siRNA Details
Gene Symbol: LOC90768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027107.2 | 4 | 1490 | |||
GenBank mRNA | AK024300.1 | - | 4 | 1441 | ||
BC033172.1 | - | 1 | 942 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG480911.1 | - | 3 | 855 | Chr. 4 - 182142396 - 182143776 [-] on Build GRCh38 |
