siRNA Details
Gene Symbol: LOC90784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026984.1 | 1 | 505 | |||
GenBank mRNA | AK001612.1 | - | 1 | 149 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC90784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026984.1 | 1 | 505 | |||
GenBank mRNA | AK001612.1 | - | 1 | 149 |