siRNA Details
Gene Symbol: LINC00092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024129.1 | 2 | 582 | |||
GenBank mRNA | BC043559.1 | - | 2 | 582 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DR006960.1 | - | 2 | 455 | Chr. 9 - 96020639 - 96021628 [-] on Build GRCh38 |
