siRNA Details
Gene Symbol: CHKB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021492.2 | 2 | 334 | |||
GenBank mRNA | BE393070.1 | - | 2 | 233 | ||
BJ989458.1 | - | 2 | 215 | |||
CR456603.1 | - | 2 | 334 | |||
CR980518.1 | - | 2 | 238 |
