siRNA Details
Gene Symbol: MZF1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027334.2 | 6 | 1998 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MZF1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027334.2 | 6 | 1998 |