siRNA Details
Gene Symbol: DKFZP586I1420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002186.1 | 1 | 213 | |||
GenBank mRNA | AL050378.1 | - | 1 | 213 | ||
BC073840.1 | - | 1 | 397 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DKFZP586I1420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002186.1 | 1 | 213 | |||
GenBank mRNA | AL050378.1 | - | 1 | 213 | ||
BC073840.1 | - | 1 | 397 |