siRNA Details
Gene Symbol: LINC00879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015400.1 | 2 | 137 | |||
GenBank mRNA | BC038721.1 | - | 2 | 140 | ||
BC048119.1 | - | 2 | 138 | |||
BC104171.1 | - | 2 | 56 | |||
BC104172.1 | - | 2 | 89 | |||
BC115359.1 | - | 2 | 56 | |||
BC115360.1 | - | 2 | 89 |
