siRNA Details
Gene Symbol: LINC00957 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015401.2 | 2 | 288 | |||
NR_072981.1 | 2 | 288 | ||||
NR_072982.1 | 2 | 154 | ||||
GenBank mRNA | AK092709.1 | - | 2 | 230 | ||
BC081571.1 | - | 2 | 166 | |||
DA066910.1 | - | 2 | 285 | |||
DA204840.1 | - | 2 | 125 | |||
DA212423.1 | - | 2 | 238 | |||
DA293391.1 | - | 2 | 123 | |||
DA406714.1 | - | 2 | 104 | |||
DA776150.1 | - | 2 | 288 |
