siRNA Details
Gene Symbol: CIRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023312.2 | 6 | 2684 | |||
NR_023313.2 | 5 | 2516 | ||||
GenBank mRNA | AK094781.1 | - | 5 | 2450 | ||
AK095781.1 | - | 4 | 2743 | |||
AK128423.1 | - | 5 | 2537 | |||
AL133636.1 | - | 1 | 893 | |||
BI819825.1 | - | 1 | 635 |
