siRNA Details
Gene Symbol: CIRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023312.2 | 6 | 2348 | |||
NR_023313.2 | 5 | 2180 | ||||
XM_011527668.1 | XP_011525970.1 | 7 | 781 | |||
GenBank mRNA | AK094781.1 | - | 5 | 2113 | ||
AK095781.1 | - | 4 | 2407 | |||
AK128423.1 | - | 5 | 2201 | |||
AL133636.1 | - | 1 | 557 | |||
BI819825.1 | - | 1 | 300 | |||
CX785176.1 | - | 1 | 519 |
