siRNA Details
Gene Symbol: LINC01018 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024423.2 | 5 | 867 | |||
NR_024424.2 | 3 | 881 | ||||
GenBank mRNA | AK125283.1 | - | 5 | 834 | ||
BC043253.2 | - | 3 | 570 | |||
BC106918.2 | - | 3 | 865 | |||
BC106919.1 | - | 3 | 868 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI560877.1 | - | 3 | 575 | Chr. 5 - 6583592 - 6586354 [+] on Build GRCh38 |
