siRNA Details
Gene Symbol: LOC100133050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027503.1 | 2 | 104 | |||
GenBank mRNA | BI830432.1 | - | 2 | 104 | ||
BQ109458.1 | - | 2 | 104 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100133050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027503.1 | 2 | 104 | |||
GenBank mRNA | BI830432.1 | - | 2 | 104 | ||
BQ109458.1 | - | 2 | 104 |