siRNA Details
Gene Symbol: LOC91450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026998.1 | 1 | 253 | |||
GenBank mRNA | AL109712.1 | - | 1 | 253 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX104738.1 | - | 1 | 250 | Chr. 15 - 77993481 - 77994222 [-] on Build GRCh38 |
