siRNA Details
Gene Symbol: LINC00935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029448.1 | 1 | 73 | |||
NR_029449.1 | 1 | 73 | ||||
GenBank mRNA | AK131005.1 | - | 1 | 73 | ||
BC036043.1 | - | 1 | 48 | |||
BC054859.1 | - | 1 | 55 | |||
BC146870.1 | - | 1 | 55 | |||
BC146877.1 | - | 1 | 55 |
