siRNA Details
Gene Symbol: DKFZP434H168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026889.1 | 1 | 1605 | |||
GenBank mRNA | AK124129.1 | - | 1 | 1602 | ||
AL117580.1 | - | 1 | 972 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DKFZP434H168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026889.1 | 1 | 1605 | |||
GenBank mRNA | AK124129.1 | - | 1 | 1602 | ||
AL117580.1 | - | 1 | 972 |