siRNA Details
Gene Symbol: DKFZP434H168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026889.1 | 1 | 220 | |||
GenBank mRNA | AK124129.1 | - | 1 | 217 | ||
CD171837.1 | - | 1 | 220 |

More Information
Other Species Targeted

