siRNA Details
Gene Symbol: DKFZP434L187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026771.1 | 3 | 1116 | |||
GenBank mRNA | AL117445.1 | - | 2 | 1006 | ||
BC130408.1 | - | 3 | 1116 | |||
BC130410.1 | - | 3 | 1116 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DKFZP434L187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026771.1 | 3 | 1116 | |||
GenBank mRNA | AL117445.1 | - | 2 | 1006 | ||
BC130408.1 | - | 3 | 1116 | |||
BC130410.1 | - | 3 | 1116 |