siRNA Details
Gene Symbol: TOLLIP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029409.1 | 1 | 761 | |||
GenBank mRNA | BC042808.1 | - | 1 | 642 | ||
BC066355.1 | - | 1 | 566 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TOLLIP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029409.1 | 1 | 761 | |||
GenBank mRNA | BC042808.1 | - | 1 | 642 | ||
BC066355.1 | - | 1 | 566 |