siRNA Details
Gene Symbol: ADAM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001569.3 | 5 | 523 | |||
NR_024106.2 | 5 | 523 | ||||
NR_024107.2 | 5 | 523 | ||||
NR_073423.1 | 5 | 523 | ||||
GenBank mRNA | AK308250.1 | - | 5 | 387 | ||
BG717260.1 | - | 5 | 528 | |||
DB447356.1 | - | 5 | 408 |
