siRNA Details
Gene Symbol: ADAM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001569.3 | 10, 11 | 1144 | |||
NR_024106.2 | 10, 11 | 1144 | ||||
NR_024107.2 | 10, 11 | 1144 | ||||
NR_073423.1 | 10, 11 | 1144 | ||||
GenBank mRNA | AK302269.1 | - | 7, 8 | 691 | ||
AK308250.1 | - | 10, 11 | 1008 |
