siRNA Details
Gene Symbol: KIAA0087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022006.1 | 2 | 1163 | |||
GenBank mRNA | BC128257.1 | - | 2 | 956 | ||
D42038.1 | - | 2 | 1126 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KIAA0087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022006.1 | 2 | 1163 | |||
GenBank mRNA | BC128257.1 | - | 2 | 956 | ||
D42038.1 | - | 2 | 1126 |