siRNA Details
Gene Symbol: PIP5K1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027712.1 | 1 | 3417 | |||
GenBank mRNA | BC028580.2 | - | 1 | 3417 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PIP5K1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027712.1 | 1 | 3417 | |||
GenBank mRNA | BC028580.2 | - | 1 | 3417 |