siRNA Details
Gene Symbol: DGCR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026651.1 | 1 | 134 | |||
GenBank mRNA | BU584104.1 | - | 1 | 133 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DGCR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026651.1 | 1 | 134 | |||
GenBank mRNA | BU584104.1 | - | 1 | 133 |