siRNA Details
Gene Symbol: DGCR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026651.1 | 1 | 475 | |||
GenBank mRNA | L77559.1 | - | 1 | 314 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM727772.1 | - | 1 | 34 | Chr. 22 - 19023065 - 19023494 [+] on Build GRCh38 |
