siRNA Details
Gene Symbol: LINC01558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026773.1 | 3 | 673 | |||
GenBank mRNA | AB016901.1 | - | 3 | 685 | ||
BC027853.1 | - | 3 | 696 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI771211.1 | - | 3 | 701 | Chr. 6 - 167785386 - 167796859 [-] on Build GRCh38 |
