siRNA Details
Gene Symbol: LINC01257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026670.2 | 5 | 2322 | |||
GenBank mRNA | AL137487.1 | - | 5 | 2322 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026670.2 | 5 | 2322 | |||
GenBank mRNA | AL137487.1 | - | 5 | 2322 |