siRNA Details
Gene Symbol: LINC01420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015367.2 | 3 | 634 | |||
GenBank mRNA | BC048131.1 | - | 3 | 660 | ||
BC053960.1 | - | 3 | 660 | |||
BC053962.1 | - | 3 | 660 | |||
BC062451.1 | - | 3 | 638 | |||
BX537532.1 | - | 3 | 667 |
