siRNA Details
Gene Symbol: SDHAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003264.2 | 16, 17 | 2321 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SDHAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003264.2 | 16, 17 | 2321 |