siRNA Details
Gene Symbol: PAXIP1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024476.1 | 1 | 113 | |||
NR_024477.1 | 1 | 113 | ||||
GenBank mRNA | AK056609.1 | - | 1 | 103 | ||
AK311721.1 | - | 1 | 113 | |||
BC033053.1 | - | 1 | 139 | |||
BC050402.1 | - | 1 | 98 |
